missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta 2/CD18 Antibody (10E12), Novus Biologicals™
Mouse Monoclonal Antibody
£173.00
Specifications
| Antigen | Integrin beta 2/CD18 |
|---|---|
| Clone | 10000000000000 |
| Dilution | ELISA 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin |
| Applications | ELISA |
| Classification | Monoclonal |
Description
Integrin beta 2/CD18 Monoclonal specifically detects Integrin beta 2/CD18 in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Integrin beta 2/CD18 | |
| ELISA 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| 3689 | |
| Purified recombinant fragment of CD18 expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 10000000000000 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cytokine Research, Immunology | |
| 959 beta subunit precursor)10, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1;macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, MF17, MFI7, P150 | |
| ITGB2 | |
| IgG2b | |
| Unpurified | |
| 85 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title