missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta-1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26918-1-AP-150UL
This item is not returnable.
View return policy
Description
Integrins are heterodimeric proteins made up of alpha and beta subunits. At least 18 alpha and 8 beta subunits have been described in mammals. Integrin family members are membrane receptors involved in cell adhesion and recognition in a variety of processes including embryogenesis, hemostasis, tissue repair, immune response and metastatic diffusion of tumor cells. This gene encodes a beta subunit. Multiple alternatively spliced transcript variants which encode different protein isoforms have been found for this gene.Specifications
| Integrin beta-1 | |
| Polyclonal | |
| Unconjugated | |
| ITGB1 | |
| CD29, FNRB, GPIIA, Integrin beta 1, Integrin beta-1, ITGB1, MDF2, MSK12, VLA 4 subunit beta, VLA BETA, VLAB | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3688 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P05556 | |
| ITGB1 | |
| Integrin beta-1 Fusion Protein Ag25426 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction