Learn More
Integrin beta 1/CD29 Antibody (419127) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB17783JF525
Product Code. 30165030
  • £459.00 / 0.10mL
Description
Integrin beta 1/CD29 Monoclonal antibody specifically detects Integrin beta 1/CD29 in Equine,Human,Porcine samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry,CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Integrin beta 1/CD29
Monoclonal
Janelia Fluor 525
CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA
Chinese hamster ovary cell line CHO-derived recombinant human Integrin β1/CD29, Gln21-Asp728 (predicted), Accession # P05556
0.1 mL
Cancer, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Phospho Specific, Signal Transduction, Stem Cell Markers
3688
Store at 4°C in the dark.
IgG2b
Flow Cytometry, Western Blot, Immunocytochemistry, CyTOF
419127
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Equine, Human, Pig
Purified