missing translation for 'onlineSavingsMsg'
Learn More

Integrin beta 1/CD29 Antibody (12G10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£175.00 - £412.00

Specifications

Antigen Integrin beta 1/CD29
Clone 12G10
Concentration 1.0 mg/mL
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:25-1:50, ELISA 10 ug/mL, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunocytochemistry
Applications Flow Cytometry, Immunofluorescence, Immunoprecipitation, ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18071616
View Documents
Novus Biologicals
NB100-63255-0.025MG
0.025 mg
£175.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18719463
View Documents
Novus Biologicals
NB100-63255
0.1 mg
£412.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Integrin beta 1/CD29 Monoclonal specifically detects Integrin beta 1/CD29 in Human, Mustelid samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications

Specifications

Integrin beta 1/CD29
1.0 mg/mL
Flow Cytometry, Immunofluorescence, Immunoprecipitation, ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot
Unconjugated
Mouse
Cancer, Cytokine Research, Cytoskeleton Markers, Embryonic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Signal Transduction, Stem Cell Markers
P05556
3688
Purified human beta1 integrin preparation from HT1080 fibrosarcoma cell extract.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
12G10
Western Blot 1:100-1:2000, Flow Cytometry 1:25-1:50, ELISA 10 ug/mL, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunocytochemistry
Monoclonal
Purified
RUO
Mustelid, Human
CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA
ITGB1
IgG1
Protein G purified
NB100-63255 recognises the human beta1 integrin. This has been shown to bind to a ligand induced binding site, binding being increased in the presence of fibronectin ligand. This also enhances alpha 5 - beta 1 - fibronectin interactions.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.