missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha V/CD51 Antibody (23C6) - Azide and BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Integrin alpha V/CD51 Monoclonal specifically detects Integrin alpha V/CD51 in Human, Chicken samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | Integrin alpha V/CD51 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 23C6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 10mM PBS |
| Gene Alias | antigen identified by monoclonal L230, CD51, CD51 antigen, integrin alpha-V, integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51), MSK8, Vitronectin receptor subunit alpha, VNRADKFZp686A08142 |
| Host Species | Mouse |
| Immunogen | Osteoclasts from osteoclastomas |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?