missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha V beta 6 Antibody (AvB6 53a.2), PerCP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-50449PCP
This item is not returnable.
View return policy
Description
Integrin alpha V beta 6 Monoclonal specifically detects Integrin alpha V beta 6 in Human samples. It is validated for Flow Cytometry.
Specifications
| Integrin alpha V + beta 6 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| Mouse fibroblasts retrovirally infected with cDNA to human Integrin alpha V beta 6 (3T3b6.19). Cells expressed a mouse/human Integrin alpha V beta 6 chimera. | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cellular Signaling, Stem Cells | |
| Human | |
| Purified |
| Flow Cytometry | |
| AvB6 53a.2 | |
| Flow Cytometry | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction