missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha V beta 6 Antibody (AvB6 53a.2), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-50449CL1
This item is not returnable.
View return policy
Description
Integrin alpha V beta 6 Monoclonal antibody specifically detects Integrin alpha V beta 6 in Human samples. It is validated for Flow Cytometry,Immunoprecipitation,Functional Assay,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| Integrin alpha V beta 6 | |
| Monoclonal | |
| CoraFluor 1 | |
| CD51, integrin subunit alpha V, ITGAV, MSK8, VNRA, VTNR | |
| Mouse fibroblasts retrovirally infected with cDNA to human Integrin alpha V beta 6 (3T3b6.19). Cells expressed a mouse/human Integrin alpha V beta 6 chimera. | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cellular Signaling, Stem Cells | |
| 3685 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry, Immunoprecipitation, Functional Assay, CyTOF, Immunocytochemistry/Immunofluorescence | |
| AvB6 53a.2 | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction