missing translation for 'onlineSavingsMsg'
Learn More

Integrin alpha V beta 6 Antibody (7k) - BSA Free, Novus Biologicals™

Product Code. 18676626 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.03mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18676626 0.025 mL 0.03mL
18616816 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18676626 Supplier Novus Biologicals Supplier No. NBP2504480.025ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Integrin alpha V beta 6 Monoclonal antibody specifically detects Integrin alpha V beta 6 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin alpha V beta 6
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 7k
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Flow Cytometry 20ug/mL, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100 - 1:250, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 10 μg per 100-500 μg protein lysate, CyTOF-ready
Formulation PBS
Gene Alias CD51, integrin subunit alpha V, ITGAV, MSK8, VNRA, VTNR
Host Species Rat
Immunogen 3T3b6, mouse fibroblast expressing human Integrin alpha V beta 6
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cellular Signaling, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 3685
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.