missing translation for 'onlineSavingsMsg'
Learn More

Integrin alpha V beta 3 Antibody (27.1 (VNR-1)), DyLight 550, Novus Biologicals™

Product Code. 18406617 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406617 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406617 Supplier Novus Biologicals Supplier No. NBP229719R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Integrin alpha V beta 3 Monoclonal antibody specifically detects Integrin alpha V beta 3 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin alpha V beta 3
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 27.1 (VNR-1)
Conjugate DyLight 550
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate
Gene Alias antigen identified by monoclonal L230, CD51, CD51 antigen, integrin alpha-V, integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51), MSK8, Vitronectin receptor subunit alpha, VNRADKFZp686A08142
Host Species Mouse
Immunogen This antibody is specific to Integrin Alpha VBeta 3
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3685
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.