missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha V beta 3 Antibody (27.1 (VNR-1)) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-29719
This item is not returnable.
View return policy
Description
Integrin alpha V beta 3 Monoclonal specifically detects Integrin alpha V beta 3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin (Negative).
Specifications
| Integrin alpha V beta 3 | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:1000, Immunohistochemistry-Paraffin (Negative) | |
| antigen identified by monoclonal L230, CD51, CD51 antigen, integrin alpha-V, integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51), MSK8, Vitronectin receptor subunit alpha, VNRADKFZp686A08142 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reacts with the human integrin alphaVbeta3. Shown to be specific for alphaVbeta3 by Loftus JC et al (1990) Science 249:915-918. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| 27.1 (VNR-1) | |
| Unconjugated | |
| P06756 | |
| ITGAV | |
| This antibody is specific to Integrin Alpha VBeta 3 | |
| 0.1 mg | |
| Adaptive Immunity, Angiogenesis, Cancer, Cytoskeleton Markers, Extracellular Matrix, Immunology, Signal Transduction | |
| 3685 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction