missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha E/CD103 Antibody (ITGAE/3904R), DyLight 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08855DL594
This item is not returnable.
View return policy
Description
Integrin alpha E/CD103 Monoclonal antibody specifically detects Integrin alpha E/CD103 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Integrin alpha E/CD103 | |
| Monoclonal | |
| DyLight 594 | |
| antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), MGC141996, Mucosal lymphocyte 1 antigen | |
| Recombinant human Integrin alpha E/CD103 protein fragment (aa 775-855) (exact sequence is proprietary) (Uniprot: P38570 ) | |
| 100 μg | |
| Innate Immunity | |
| 3682 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| ITGAE/3904R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction