missing translation for 'onlineSavingsMsg'
Learn More

Integrin alpha E/CD103 Antibody (Ber-ACT8) - BSA Free, Novus Biologicals™

Product Code. 18068646 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
Unit Size:
0.1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18068646 0.1 mg 0.1mg
18412211 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18068646 Supplier Novus Biologicals Supplier No. NBP197564

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Integrin alpha E/CD103 Monoclonal specifically detects Integrin alpha E/CD103 in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Flow (Cell Surface), CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin alpha E/CD103
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), CyTOF
Classification Monoclonal
Clone Ber-ACT8
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry 1-4 ug/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:50, Immunohistochemistry-Frozen 1:200, Flow (Cell Surface), CyTOF-ready
Gene Alias antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), MGC141996, Mucosal lymphocyte 1 antigen
Gene Symbols ITGAE
Host Species Mouse
Immunogen HTLV-1 induced human T cell line MAPS16
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3682
Test Specificity The mouse monoclonal antibody Ber-ACT8 recognizes an extracellular epitope of CD103 (alpha E integrin), a type I transmembrane glycoprotein primarily found on intestinal intraepithelial lymphocytes. HLDA V; WS Code A067
Target Species Human, Primate
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.