missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha E/CD103 Antibody (AP-MAB0828) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
£147.00
Specifications
| Antigen | Integrin alpha E/CD103 |
|---|---|
| Clone | AP-MAB0828 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, CyTOF-ready |
| Applications | Flow Cytometry |
Description
Integrin alpha E/CD103 Monoclonal specifically detects Integrin alpha E/CD103 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, CyTOF-ready.Specifications
| Integrin alpha E/CD103 | |
| 1 mg/mL | |
| Flow Cytometry | |
| Unconjugated | |
| Rat | |
| Innate Immunity | |
| PBS with No Preservative | |
| 3682 | |
| Mouse mammary tumor cells | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| AP-MAB0828 | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), MGC141996, Mucosal lymphocyte 1 antigen | |
| ITGAE | |
| IgG2 | |
| Protein A or G purified | |
| Integrin alpha IEL chain |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title