missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha E/CD103 Antibody (2G5.1), Novus Biologicals™
Mouse Monoclonal Antibody
£160.00 - £386.00
Specifications
| Antigen | Integrin alpha E/CD103 |
|---|---|
| Clone | 2G5.1 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:50-1:100 |
| Applications | Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18003184
|
Novus Biologicals
NB100-65271-0.025MG |
0.025 mg |
£160.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18050146
|
Novus Biologicals
NB100-65271 |
0.1 mg |
£386.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Integrin alpha E/CD103 Monoclonal specifically detects Integrin alpha E/CD103 in Human, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| Integrin alpha E/CD103 | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Innate Immunity | |
| P38570 | |
| 3682 | |
| Human Intestinal lymphocyte | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 2G5.1 | |
| Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:50-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Primate | |
| antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), MGC141996, Mucosal lymphocyte 1 antigen | |
| ITGAE | |
| IgG2a | |
| Protein A purified | |
| NB100-65271 recognizes the human CD103 cell surface antigen, also known as the alphaE integrin. This reacts with various subsets of human intra-epithelial lymphocytes:- 40% of lamina propria T-cells; 30% of mesenteric lymphoblasts and some lymphocytes from other mucosa. It recognizes less than 0.1% bone marrow cells and less than 0.7% PBL. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title