missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha-9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£111.18 - £274.38
Specifications
| Antigen | Integrin alpha-9 |
|---|---|
| Concentration | 0.19 mg/mL |
| Applications | Immunofluorescence, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
This gene encodes an alpha integrin. Integrins are heterodimeric integral membrane glycoproteins composed of an alpha chain and a beta chain that mediate cell-cell and cell-matrix adhesion. The protein encoded by this gene, when bound to the beta 1 chain, forms an integrin that is a receptor for VCAM1, cytotactin and osteopontin. Expression of this gene has been found to be upregulated in small cell lung cancers.Specifications
| Integrin alpha-9 | |
| Immunofluorescence, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| B8JK39, Q13797 | |
| 104099, 3680, 685004 | |
| Integrin alpha-9 Fusion Protein Ag6126 | |
| Primary | |
| -20°C | |
| ITGA9 |
| 0.19 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| ALPHA RLC, Integrin alpha 9, Integrin alpha RLC, integrin, alpha 9, ITGA4L, ITGA9, RLC | |
| ITGA9 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title