Learn More
Integrin alpha 8 Antibody (CL7304) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76482JF646
Product Code. 30296115
  • £558.00 / 0.10mL
Description
Integrin alpha 8 Monoclonal antibody specifically detects Integrin alpha 8 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Integrin alpha 8
Monoclonal
Janelia Fluor 646
integrin alpha-8, integrin, alpha 8
This antibody was developed using a recombinant protein derived from P53708, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL7304
50mM Sodium Borate
Mouse
Protein A purified
RUO
8516
Store at 4⪚C in the dark.
IgG1