Learn More
Integrin alpha 6/CD49f Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-85747
This item is not returnable.
View return policy
Description
Integrin alpha 6/CD49f Polyclonal antibody specifically detects Integrin alpha 6/CD49f in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin).
Specifications
| Immunohistochemistry, Immunoprecipitation | |
| ITGA6 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:APYDDLGKVFIYHGSANGINTKPTQVLKGISPYFGYSIAGNMDLDRNSYPDVAVGSLSDSVTIFRSRPVINIQKTITVTPNRIDLRQKTACGAPSGICLQVKSCFEYTANPAGYNPSISIVGTLEAEKERRKSGLSS | |
| 0.1 mL |
| CD49 antigen-like family member F, CD49f, CD49f antigen, DKFZp686J01244, FLJ18737, integrin alpha-6, integrin alpha6B, integrin, alpha 6, ITGA6B, VLA-6 | |
| Rabbit | |
| 127 kDa | |
| RUO |