Learn More
Integrin alpha 6/CD49f Antibody (CL6957) [Alexa Fluor™ 647], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76523AF647
Product Code. 30295523
  • £558.00 / 0.10mL
Description
Integrin alpha 6/CD49f Monoclonal antibody specifically detects Integrin alpha 6/CD49f in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Integrin alpha 6/CD49f
Monoclonal
Alexa Fluor 647
CD49 antigen-like family member F, CD49f, CD49f antigen, DKFZp686J01244, FLJ18737, integrin alpha-6, integrin alpha6B, integrin, alpha 6, ITGA6B, VLA-6
This antibody was developed using a recombinant protein derived from P23229, with the exact immunogen sequence remaining proprietary.
0.1 mL
Signal Transduction
3655
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL6957
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified