missing translation for 'onlineSavingsMsg'
Learn More

Integrin alpha 5/CD49e Antibody (SAM1), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18338203 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338203 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338203 Supplier Novus Biologicals Supplier No. NBP318529JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Integrin alpha 5/CD49e Monoclonal antibody specifically detects Integrin alpha 5/CD49e in Human, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin alpha 5/CD49e
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone SAM1
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias CD49 antigen-like family member E, CD49e, CD49e antigen, Fibronectin receptor subunit alpha, fibronectin receptor, alpha subunit, FNRAVLA5A, integrin alpha-5, Integrin alpha-F, integrin, alpha 5 (fibronectin receptor, alpha polypeptide), very late activation protein 5, alpha subunit, VLA-5
Host Species Mouse
Immunogen U937 cells
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline CD Markers, Cell Biology, Cytoskeleton Markers, GPCR, Immunology, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 3678
Target Species Human, Primate
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.