missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 5/CD49e Antibody (CL6940) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44402
This item is not returnable.
View return policy
Description
Integrin alpha 5/CD49e Monoclonal antibody specifically detects Integrin alpha 5/CD49e in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry,KnockDown
Specifications
| Integrin alpha 5/CD49e | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 μg/mL, Immunohistochemistry-Paraffin 1:5000 - 1:10000 | |
| CD49 antigen-like family member E, CD49e, CD49e antigen, Fibronectin receptor subunit alpha, fibronectin receptor, alpha subunit, FNRAVLA5A, integrin alpha-5, Integrin alpha-F, integrin, alpha 5 (fibronectin receptor, alpha polypeptide), very late activation protein 5, alpha subunit, VLA-5 | |
| This antibody was generated using a recombinant protein sequence of P08648, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| CD Markers, Cell Biology, Cytoskeleton Markers, GPCR, Immunology, Signal Transduction, Stem Cells | |
| 3678 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry, KnockDown | |
| CL6940 | |
| Unconjugated | |
| Lyophilized in 5% Trehalose. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
| Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction