missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 5 beta 1 Antibody (M200 (Volociximab)) - Azide and BSA Free, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP2-75909-0.2MG
This item is not returnable.
View return policy
Description
Integrin alpha 5 beta 1 Monoclonal specifically detects Integrin alpha 5 beta 1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Block/Neutralize.
Specifications
| Integrin alpha 5 beta 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.02% Proclin 300 | |
| Human | |
| 0.2 mg | |
| The antibody binds to human (alpha)5(beta)1 integrin with a Kd of 0.367 nM (determined by Biacore), an EC50 of 0.2 nM and an IC50 of 2.3 nM. | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Blocking Assay, Neutralization | |
| M200 (Volociximab) | |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Block/Neutralize | |
| ITGA5 | |
| Heterodimeric (alpha)5(beta)1Fc fusion protein | |
| Primary | |
| Human | |
| IgG4 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction