missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Integrin alpha 5 beta 1 Antibody (BMS patent anti-Integrin alpha5beta1) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28423-50ug
This item is not returnable.
View return policy
Description
Integrin alpha 5 beta 1 Monoclonal antibody specifically detects Integrin alpha 5 beta 1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Integrin alpha 5 beta 1 | |
| Monoclonal | |
| Unconjugated | |
| CD49 antigen-like family member E, CD49e, Fibronectin receptor subunit alpha, fibronectin receptor, alpha polypeptide, fibronectin receptor, alpha subunit, FNRA, integrin alpha-5, Integrin alpha-F, integrin, alpha 5 (fibronectin receptor, alpha polypeptide), ITGA5/SLC9B1 fusion, very late activation protein 5, alpha subunit, VLA-5, VLA5A | |
| Integrin a5b1 (ITGA5 & ITGB1) | |
| 50 μg | |
| Angiogenesis, Cancer, Immunology | |
| 3678 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| BMS patent anti-Integrin alpha5beta1 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction