missing translation for 'onlineSavingsMsg'
Learn More

Integrin alpha 4/CD49d Antibody (Natalizumab) - Azide and BSA Free, Novus Biologicals™

Product Code. 18352911 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18352911 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18352911 Supplier Novus Biologicals Supplier No. NBP275916SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

Integrin alpha 4/CD49d Monoclonal specifically detects Integrin alpha 4/CD49d in Human samples. It is validated for Flow Cytometry, Block/Neutralize.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin alpha 4/CD49d
Applications Flow Cytometry, Blocking Assay, Neutralization
Classification Monoclonal
Clone Natalizumab
Conjugate Unconjugated
Dilution Flow Cytometry, Block/Neutralize
Formulation PBS with 0.02% Proclin 300
Gene Alias 269C wild type, biosimilar, biosimilars, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha
Gene Symbols ITGA4
Host Species Human
Immunogen The RAMOS cell line, a non-EBV releasing cell line, injected into mice.
Purification Method Protein A purified
Quantity 0.025 mg
Primary or Secondary Primary
Gene ID (Entrez) 3676.0
Test Specificity This antibody binds to the alpha 4 subunit of (alpha)4(beta)1 (also known as very late antigen 4 (VLA-4) or CD49d-CD29) and (alpha)4(beta)7 integrin
Target Species Human
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Form Purified
Isotype IgG4 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.