missing translation for 'onlineSavingsMsg'
Learn More

Integrin alpha 4/CD49d Antibody (21.6), DyLight 755, Novus Biologicals™

Product Code. 18331300 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18331300 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18331300 Supplier Novus Biologicals Supplier No. NBP281092IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Integrin alpha 4/CD49d Monoclonal specifically detects Integrin alpha 4/CD49d in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin alpha 4/CD49d
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 21.6
Conjugate DyLight 755
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias 269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha
Gene Symbols ITGA4
Host Species Rabbit
Immunogen Integrin alpha 4/CD49d
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones, Microautophagy, Neuronal Cell Markers, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3676
Test Specificity This antibody binds to human Integrin alpha 4/CD49d
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.