missing translation for 'onlineSavingsMsg'
Learn More

Integrin alpha 3B Antibody (54B3), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18411758 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411758 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411758 Supplier Novus Biologicals Supplier No. NBP197732AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Integrin alpha 3B Monoclonal antibody specifically detects Integrin alpha 3B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin alpha 3B
Applications Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 54B3
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias AA407068, CD49C, GAPB3, integrin alpha 3
Host Species Mouse
Immunogen Derived by fusion of SP2/0 Mouse myeloma cells with spleen cells from a BALB/c Mouse immunized with a synthetic peptide corresponding to a 32 amino acid stretch in the cytoplasmic domain of Integrin alpha 3Bincluding an appending N-terminal cysteine (CTRYYQIMPKYHAVRIREEERYPPPGSTLPTKK) coupled to keyhole limpet hemocyanin.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3675
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.