missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 3/CD49c Antibody (CL6937) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76515APC
This item is not returnable.
View return policy
Description
Integrin alpha 3/CD49c Monoclonal antibody specifically detects Integrin alpha 3/CD49c in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
Specifications
| Integrin alpha 3/CD49c | |
| Monoclonal | |
| APC | |
| antigen identified by monoclonal J143, CD49 antigen-like family member C, CD49c, CD49c antigen, FLJ34631, FLJ34704, FRP-2, Galactoprotein B3, GAP-B3, GAPB3CD49C, integrin alpha-3, integrin, alpha 3 (antigen CD49C, alpha 3 subunit of VLA-3 receptor), MSK18, VCA-2, very late activation protein 3 receptor, alpha-3 subunit, VL3A, VLA-3 subunit alpha, VLA3a | |
| This antibody was developed using a recombinant protein derived from P26006, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown | |
| CL6937 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 3675 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction