missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 3/CD49c Antibody (ASC-1), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62200G
This item is not returnable.
View return policy
Description
Integrin alpha 3/CD49c Monoclonal specifically detects Integrin alpha 3/CD49c in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| Integrin alpha 3/CD49c | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ITGA3 | |
| Human SSC-9 cell line (squamous cell carcinoma) | |
| 0.1 mL | |
| 3675 | |
| Human | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| ASC-1 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
| antigen identified by monoclonal J143, CD49 antigen-like family member C, CD49c, CD49c antigen, FLJ34631, FLJ34704, FRP-2, Galactoprotein B3, GAP-B3, GAPB3CD49C, integrin alpha-3, integrin, alpha 3 (antigen CD49C, alpha 3 subunit of VLA-3 receptor), MSK18, VCA-2, very late activation protein 3 receptor, alpha-3 subunit, VL3A, VLA-3 subunit alpha, VLA3a | |
| Mouse | |
| Protein A purified | |
| Primary | |
| The antibody HTF-14 recognizes an epitope located in the N-terminal domain of human serum transferrin, a 77 kDa single polypeptide chain glycoprotein (member of the iron binding family of proteins). It is synthesised in the liver and consists of two domains each having a high affinity reversible binding site for Fe3+. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction