missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Integrin alpha 2/CD49b Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 2 publications
POA
Specifications
Antigen | Integrin alpha 2/CD49b |
---|---|
Clone | 430907 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
Integrin alpha 2/CD49b Monoclonal specifically detects Integrin alpha 2/CD49b in Human samples. It is validated for Western Blot.Specifications
Integrin alpha 2/CD49b | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Human | |
P17301 | |
3673 | |
Chinese hamster ovary cell line CHO-derived recombinant human Integrin alpha 2/CD49b Tyr30-Thr1132 Accession # P17301 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Integrin alpha 2/CD49b in direct ELISAs and Western blots. In direct ELISAs and Western blots, 50-100% cross-reactivity with recombinant mouse (rm) Integrin alpha 2 is observed and no cross-reactivity with recombinant human (rh) Integrin alpha 1, alpha 3, alpha 5, alpha 6, alpha 8, alpha 9, alpha 10, alpha 11, alpha L, alpha M, or alpha V is observed. |
430907 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
alpha-2 subunit, CD49b, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), ITGA2, VLA 2, VLA-2 alpha | |
ITGA2 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only