missing translation for 'onlineSavingsMsg'
Learn More

Integrin alpha 2/CD49b Antibody (430907) [Janelia Fluor™ 549], Novus Biologicals™

Product Code. 30734773 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734773 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734773 Supplier Novus Biologicals Supplier No. FAB12331I

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Integrin alpha 2/CD49b Monoclonal antibody specifically detects Integrin alpha 2/CD49b in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin alpha 2/CD49b
Applications Western Blot
Classification Monoclonal
Clone 430907
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias alpha-2 subunit, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), VLA 2
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Integrin alpha 2/CD49b, Tyr30-Thr1132, Accession # P17301
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3673
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.