Learn More
Integrin alpha 10 Antibody (885501), mFluor Violet 450 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB9757MFV450
This item is not returnable.
View return policy
Description
Integrin alpha 10 Monoclonal antibody specifically detects Integrin alpha 10 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Integrin alpha 10 | |
| CyTOF | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha 10 beta 1, Phe23-Thr1119 (Integrin alpha 10) and Gln21-Asp728 (Integrin beta 1), Accession # NP_001289400.1 (Integrin alpha 10) and P09055 (Integrin beta 1) | |
| 0.1 mL |