missing translation for 'onlineSavingsMsg'
Learn More

MEDICOM Integra PVC High Risk Gloves

Product Code. 31781339 Shop All Medicom Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
Pack of 200
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
31781339 Large Pack of 200
31781393 Medium Pack of 200
31781368 Small Pack of 200
31781420 X-Large Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31781339 Supplier MEDICOM Supplier No. 30.G52N/L
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The Integra PVC High Risk glove is Category III PPE, effectively protecting against mechanical and chemical risks thanks to its watertight 400 mm cuff.

  • The PVC glove is designed to protect hands against mechanical and chemical hazards
  • The glove is made of white PVC with a long translucent PVC cuff 400 mm long
  • The glove is waterproof and airtight
  • It is particularly recommended for use in the chemical industry, maintenance and heavy industry: agriculture, construction, waste collection, laundry and industrial cleaning

Specifications

Applications Industrial
Color White
Latex Free Yes
Material PVC
Product Type Disposable Gloves
Size Large
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.