missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INT7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584902
This item is not returnable.
View return policy
Description
Immunogen sequence: GDGMLGDLME LYKVIGRSAT DKQQELLVSL ATVIFVASQK ALSVESKAVI KQQLESVSNG WTVYRIARQA SRMGNHDMAK ELYQSLLTQV A Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 95%.
This gene encodes a subunit of the integrator complex. The integrator complex associates with the C-terminal domain of RNA polymerase II and mediates 3'-end processing of the small nuclear RNAs U1 and U2. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
| INT7 | |
| Polyclonal | |
| Unconjugated | |
| INTS7 | |
| 5930412E23Rik; C1orf73; DKFZP434B168; Int7; Integrator complex subunit 7; Ints7; RGD1308014 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25896 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NVH2 | |
| INTS7 | |
| Recombinant protein corresponding to Human INTS7. Recombinant protein control fragment (Product #RP-108599). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction