missing translation for 'onlineSavingsMsg'
Learn More

INT4 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB100-60660H

Product Code. 18551822

  • £492.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

INT4 Polyclonal antibody specifically detects INT4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

INT4
Polyclonal
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
INT4, integrator complex subunit 4, MGC16733, MST093, MSTP093
The immunogen recognized by this antibody maps to a region between residue 913 and 963 of integrator complex subunit 4 using the numbering given in entry NP_291025.3 (GeneID 92105).
0.1 mL
Primary
Human, Mouse
Purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
HRP
PBS
Rabbit
Immunogen affinity purified
RUO
92105
Store at 4C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.