missing translation for 'onlineSavingsMsg'
Learn More

INT-777, MedChemExpress

Product Code. p-200101655
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16452817 50 mg 50mg
16449287 5 mg 5mg
16446738 10 mg 10mg
16416876 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16452817 Supplier Medchem Express Supplier No. HY15677

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress INT-777 is a potent TGR5 agonist with an EC50 of 0.82 μM.

INT-777 is a potent TGR5 agonist with an EC50 of 0.82 μM.

Specifications

Chemical Name or Material INT-777
CAS 1199796-29-6
Color White
Purity Grade Notes Research
Molecular Formula C27H46O5
Quantity 50 mg
Synonym S-EMCA
Solubility Information Ethanol : ≥ 50 mg/mL (110.95 mM) ∣DMSO : ≥ 31 mg/mL (68.79 mM)
SMILES O[C@@H]1CC[C@@]2(C)[C@@]([C@@H](CC)[C@@H](O)[C@]3([H])[C@]2([H])C[C@H](O)[C@@]4(C)[C@@]3([H])CC[C@@H]4[C@H](C)C[C@H](C)C(O)=O)([H])C1
Molecular Weight (g/mol) 450.65
Formula Weight 450.65
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.