missing translation for 'onlineSavingsMsg'
Learn More

INT-767, MedChemExpress

Product Code. 16477887
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16477887 5 mg 5mg
16471387 50 mg 50mg
16435168 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16477887 Supplier Medchem Express Supplier No. HY12434
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress INT-767 is a dual farnesoid X receptor (FXR)/TGR5 agonist with mean EC50s of 30 and 630 nM, respectively.

INT-767 is a dual farnesoid X receptor (FXR)/TGR5 agonist with mean EC50s of 30 and 630 nM, respectively.

Specifications

Chemical Name or Material INT-767
CAS 1000403-03-1
Color White
Purity Grade Notes Research
Molecular Formula C25H43NaO6S
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (202.16 mM) ∣H2O : 100 mg/mL (202.16 mM; Need ultrasonic)
SMILES C[C@H](CCOS(=O)(O[Na])=O)[C@@]1([H])CC[C@@]2([H])[C@]3([H])[C@H](O)[C@H](CC)[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])CC[C@@]21C
Molecular Weight (g/mol) 494.66
Formula Weight 494.66
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.