missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulysin/IDE Rabbit anti-Human, Mouse, Rat, Clone: 8O8F3, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Insulysin/IDE Monoclonal antibody specifically detects Insulysin/IDE in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Insulysin/IDE |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 8O8F3 |
| Conjugate | Unconjugated |
| Formulation | PBS, 0.05% BSA, 50% glycerol, pH7.3 |
| Gene Alias | Abeta-degrading protease, EC 3.4.24, EC 3.4.24.56, FLJ35968, Insulin protease, insulinase, insulin-degrading enzyme, Insulysin |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Insulysin/IDE (P14735). |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?