missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin R/CD220, Rabbit anti-Human, Clone: 83-7, Novus Biologicals™
Rabbit Monoclonal Antibody
£207.00 - £463.00
Specifications
| Antigen | Insulin R/CD220 |
|---|---|
| Clone | 83-7 |
| Concentration | 1 mg/mL |
| Dilution | ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin |
| Applications | ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18329220
|
Novus Biologicals
NBP2-81039-0.025MG |
0.025 mg |
£207.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18361580
|
Novus Biologicals
NBP2-81039-0.2MG |
0.2 mg |
£463.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Insulin R/CD220 Monoclonal specifically detects Insulin R/CD220 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Insulin R/CD220 | |
| 1 mg/mL | |
| ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Diabetes Research, Lipid and Metabolism, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| PBS | |
| 3643 | |
| Epitope within amino acids 140-301 (the cysteine rich region) of the extracellular domain of the human Insulin R/CD220. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 83-7 | |
| ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, insulin receptor, IR | |
| INSR | |
| IgG κ | |
| Protein A purified | |
| Recognises the extracellular domain of the Insulin R/CD220. Amino acids 469 - 592 (exon 7 and 8) of Insulin R/CD220. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title