missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin R/CD220 Mouse anti-Human, Clone: INSR/1661, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07512-100ug
This item is not returnable.
View return policy
Description
Insulin R/CD220 Monoclonal specifically detects Insulin R/CD220 in Human samples. It is validated for ELISA.
Specifications
| Insulin R/CD220 | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA 2 to 4 μg/mL | |
| CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, insulin receptor, IR | |
| Recombinant fragment of extracellular domain of human Insulin R/CD220 (exact sequence is proprietary) | |
| 100 μg | |
| Diabetes Research, Lipid and Metabolism, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 3643 | |
| Store at 4°C. | |
| IgG1 κ |
| ELISA | |
| INSR/1661 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction