missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin R/CD220 Antibody (INSR/1661) - Azide and BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Insulin R/CD220 Monoclonal specifically detects Insulin R/CD220 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | Insulin R/CD220 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | INSR/1661 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | ELISA : 2-4 ug/mL |
| Formulation | 10mM PBS |
| Gene Alias | CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, insulin receptor, IR |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
¿Detecta una oportunidad de mejora?