missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin R/CD220 Antibody (83-14), mFluor Violet 450 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Insulin R/CD220 Monoclonal antibody specifically detects Insulin R/CD220 in Human, Porcine, Bovine, Sheep samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | Insulin R/CD220 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 83-14 |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, insulin receptor, IR |
| Host Species | Rabbit |
| Immunogen | This antibody was raised by immunizing mice with IM-9 lymphocytes followed by purified insulin receptor. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?