missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Insulin(cattle)

Product Code. 16474046
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16474046 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16474046 Supplier MedChemExpress Supplier No. HYP1156

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Insulin cattle (Insulin from bovine pancreas) is a two-chain polypeptide hormone produced in vivo in the pancreatic β cells. Insulin cattle has often been used as growth supplement in culturing cells.

Specifications

CAS 11070-73-8
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
For Use With (Application) Metabolism-sugar/lipid metabolism
Molecular Weight (g/mol) 5733.49 g/mol
Product Type Peptides
Quantity 25 mg
Shipping Condition Room temperature
Solubility H2O : 10 mg/mL (1.74 mM∣ultrasonic and warming and adjust pH to 3 with HCl and heat to 60°C)
Synonym Insulin from bovine pancreas
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.