missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin Antibody (rIRDN/8546), Novus Biologicals™
Mouse Monoclonal Antibody
£195.00 - £385.00
Specifications
| Antigen | Insulin |
|---|---|
| Clone | rIRDN/8546 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 0.1-0.2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18600845
|
Novus Biologicals
NBP3-20522-100ug |
100 μg |
£385.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18683784
|
Novus Biologicals
NBP3-20522-20ug |
20 μg |
£195.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Insulin Monoclonal antibody specifically detects Insulin in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Insulin | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Stem Cell Markers | |
| 10mM PBS with 0.05% BSA | |
| 3630 | |
| IgG2a κ | |
| Protein A or G purified |
| rIRDN/8546 | |
| Immunohistochemistry-Paraffin 0.1-0.2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| Recombinant full-length human Insulin protein | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title