missing translation for 'onlineSavingsMsg'
Learn More

Insulin Antibody (E2-E3 + 2D11-H5 (same as INS04 + INS05)) - Azide and BSA Free, Novus Biologicals™

Product Code. 18402422 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402422 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402422 Supplier Novus Biologicals Supplier No. NBP2346120.1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Insulin Monoclonal antibody specifically detects Insulin in Human, Mouse, Porcine, Bovine, Rabbit samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Insulin
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone E2-E3 + 2D11-H5 (same as INS04 + INS05)
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation 10 mM PBS
Gene Alias IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Host Species Mouse
Immunogen Purified pig insulin, conjugated to KLH (E2-E3 & 2D11-H5) (Uniprot: P01308)
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Diabetes Research, Immune System Diseases, Immunology, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3630
Target Species Human, Mouse, Pig, Bovine, Rabbit
Content And Storage Store at -20°C to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.