missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ PVC-Coated Triple Insulated Gloves - Reinforced Safety Cuff

Product Code. 12970775
Click to view available options
:
Large
This item is not returnable. View return policy

Product Code. 12970775

Brand: SHOWA™ 7610

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Three layers of foam insulation provide protection against chemicals, and cold weather. PVC-coating has a rough finish for enhanced grip capabilities in wet or dry conditions.

  • Three layers of foam insulation between a layer of cotton-jersey and cotton interlock for increased cold-weather protection
  • Low-temperature compound remains flexible, even in sub-zero temperatures
  • Reinforced safety cuff extends coverage to wrist
  • PVC-coating provides abrasion and chemical resistance
  • Rough finish enhances grip capabilities in wet and dry conditions

Specifications

Cuff Material Reinforced Cotton
Coating Material PVC
Color Orange
Exterior Finish Fully Textured
Hand Specific
Material Cotton
Size Large
For Use With (Application) Signalers, Digging, Utility Work, and Heavy Equipment Handling
Glove Cuff Style Safety
Liner Material Cotton Jersey
Series 76
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.