missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Instrument Control Terbium TR-FRET Kit
Click to view available options
Quantity:
1 mL kit
Unit Size:
Each
Description
The Instrument Control Terbium TR-FRET Kit allows for the rapid assessment of instrument setup for terbium (Tb)-based TR-FRET assays. LanthaScreen™ and Premo™ assays require specific instrument settings. Proper instrument setup is critical to experimental success. This kit contains one tube each of 1x high and low controls which may also be used as positive control samples when running Tb TR-FRET assays.
The Instrument Control Terbium TR-FRET Kit Allows You to:
Set up instruments for Tb TR-FRET assays quickly
Check instrument Tb TR-FRET settings
Read a high and low Tb TR-FRET signal control on each assay plate
Determine if Your Instrument is Set Up to Read Tb TR-FRET Assays
Setting up your instrument to read Tb TR-FRET is the first step in running a successful Tb TR-FRET assay. This kit allows you to quickly determine if your instrument is set up properly to read Tb TR-FRET assays. Simply add a small volume into separate wells, place the plate into the instrument and read. If the ratio of high to low control signal is below 2 the instrument may not be setup properly. If the ratio of high to low control signal is between 2 and 4 you can proceed with a Tb TR-FRET.
Use the Kit as a Positive Control in Tb TR-FRET Assays
When running Tb TR-FRET assays, add a small volume of the high and low controls to separate wells, on your assay plate. Place the plate into the instrument and read, according to instructions. A high to low control signal ratio of between 2 and 4 offers you the confidence that the instrument is performing well.
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
The Instrument Control Terbium TR-FRET Kit Allows You to:
Determine if Your Instrument is Set Up to Read Tb TR-FRET Assays
Setting up your instrument to read Tb TR-FRET is the first step in running a successful Tb TR-FRET assay. This kit allows you to quickly determine if your instrument is set up properly to read Tb TR-FRET assays. Simply add a small volume into separate wells, place the plate into the instrument and read. If the ratio of high to low control signal is below 2 the instrument may not be setup properly. If the ratio of high to low control signal is between 2 and 4 you can proceed with a Tb TR-FRET.
Use the Kit as a Positive Control in Tb TR-FRET Assays
When running Tb TR-FRET assays, add a small volume of the high and low controls to separate wells, on your assay plate. Place the plate into the instrument and read, according to instructions. A high to low control signal ratio of between 2 and 4 offers you the confidence that the instrument is performing well.
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
Specifications
Specifications
| Product Line | LanthaScreen |
| Shipping Condition | Wet Ice |
| Color | Green |
| For Use With (Equipment) | HTS Reader, Microplate Reader |
| Quantity | 1 mL kit |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction