missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ 3500xL Dx Genetic Analyzer Installation Kit
Shop All Applied Biosystems ProductsDescription
Includes
Installation kit comes with a 50cm 24-capillary array, POP-6™ and POP-7™ polymers, Big Dye™ Terminator v1.1 and v3.1 Sequencing Standards, and a fragment analysis reagent including DS-33 GeneScan™ installation kit.
The 3500xL Dx Genetic Analyzer Installation Kit includes all of the consumables and accessories needed to perform an instrument qualification on a 3500xL Dx Genetic Analyzer with Data Collection Software 3.
Order Info
1 Kit
Specifications
Specifications
| Content And Storage | This kit comes with a 50-cm 24-capillary array, POP-6 and POP-7 polymer, Big Dye Terminator v1.1 and v3.1 sequencing standard and fragment analysis reagent including DS-33 GS Install Kit. |
| Format | Kit |
| For Use With (Equipment) | 3500xL Dx Genetic Analyzer |
| For Use With (Application) | Sequencing |
| Label or Dye | LIZ |
| Product Type | Genetic Analyzer Installation Kit |
| Quantity | 1 Kit |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet or Dry Ice |
For in-vitro diagnostic use
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?