Learn More
Thermo Scientific™ IQOQDocE Qualification Documentation for IntelliStack Incubator Shaker (English)
Product Code. 30057378 Shop All Thermo Scientific Products
Click to view available options
:
IQOQDocE Qualification Documentation for IntelliStack Incubator Shaker (English)
Product Code. 30057378
Brand: Thermo Scientific™ IOQDOCE3510
IQOQDocE Qualification Documentation for IntelliStack Incubator Shaker (English)
  • IQ: Verifies that the installation site and equipment environment meet manufacturer-specified environmental requirements. Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order. Verifies equipment and any other accessories are assembled and installed. Configuration information for each system component gets recorded.
    • Installation of equipment is NOT included
  • OQ: Verifies important equipment functions and ensures that the equipment operates as expected by the manufacturer and in conformance to standards and requirements.
  • Key tests: Door Ajar, Power Failure, Temperature Alerts
TRUSTED_SUSTAINABILITY
Specifications
Product Type IQ/OQ Documentation