Learn More
R&D Systems™ INSTA-Blot Ovary Tissue OncoPair
Product Code. 18791154 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Membrane
Unit Size:
Each
Product Code. Quantity Unit Size
18791154 1 Membrane Each
1 options
Product Code. 18791154 Supplier R&D Systems™ Supplier No. NBP230124
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A Novus product, part of R&D Systems. A R&D Systems product, part of R&D Systems. PVDF western blot membrane containing denatured protein lysates
  • Proteomic discovery tool
  • Addresses increasing demand for inclusion of human tissue-derived products in biomarker profiling
  • Manufactured from Imgenex's extensive Human Clinical Tissue Lysate collection
  • Each blot contains 14 lanes of alternating tumor (T) and matched normal adjacent (NA) tissue lysates from seven patient donors
  • Incorporation of T and NA enables protein expression analysis of tumor and microenvironment
  • Useful in conjunction with Cell Line, Normal Tissue INSTA-BlotsTM, lysates for comprehensive antibody validation
  • Used in profile antibody reactivity, protein expression patterns across species and between tissues and cells lines
Recommended Storage
Store at room temperature
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Used in western blot for screening expression of various proteins in tumor and normal adjacent tissues from various donors
Quantity 1 Membrane
Material PVDF
Product Line Novus