missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ INSTA-Blot Multi-species Stomach, Small Intestine and Pancreas

Product Code. 18162515 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Membrane
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 18162515

Brand: Novus Biologicals™ NBP230118

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PVDF membrane containing denatured protein from cells or tissue lysates

  • Screening proteins from various human, mouse and rat tissues
  • Contains 11 human, mouse and rat tissue lysates (20μg total protein per lane) on 4-20% Tris-Glycine mini gel
  • Prepared from resolution by SDS-PAGE at 125V for 1 hour
  • Proteins are electro-blotted for 2 hours at 25V onto PVDF membrane
  • Stained with amido black for visualization of proteins

Recommended Storage

Store at room temperature

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Screening proteins from various human, mouse and rat tissues
Quantity 1 Membrane
Material PVDF
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.