missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INSRR Monoclonal Antibody (326903)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561252
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
INSRR, insulin receptor-related receptor, is a member of the insulin receptor (IR) family. INSRR does not bind insulin-like molecules, but is similar to INSR in tyrosine kinase activity. Receptor with tyrosine-protein kinase activity. Functions as a pH sensing receptor which is activated by increased extracellular pH. Activates an intracellular signaling pathway that involves IRS1 and AKT1/PKB.
Specifications
| INSRR | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P14616 | |
| Insrr | |
| Mouse myeloma cell line NS0-derived recombinant human INSRR, Leu24-Leu923. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 326903 | |
| Unconjugated | |
| Insrr | |
| Insrr; insulin receptor related receptor; insulin receptor-related protein; Insulin receptor-related protein alpha chain; Insulin receptor-related protein beta chain; insulin receptor-related receptor; IRR; IR-related receptor; Sirr | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3645 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction